Learn More
3-(Diethylboryl)pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl(3-pyridinyl)borane |
| CAS | 89878-14-8 |
| Molecular Formula | C9 H14 B N |
| Quantity | 1 g |
| Synonym | 3-(Diethylboryl)pyridine, 3-Diethylboropyridine, 3-Pyridinyldiethylborane, 3-Pyridyldiethylborane, Diethyl(3-pyridinyl)borane, Diethyl-3-pyridylborane |
| InChI Formula | InChI=1S/C9H14BN/c1-3-10(4-2)9-6-5-7-11-8-9/h5-8H,3-4H2,1-2H3 |
| SMILES | CCB(CC)c1cccnc1 |
| IUPAC Name | diethyl(pyridin-3-yl)borane |
| Molecular Weight (g/mol) | 147.0252 |
| Formula Weight | 147.1219 |