missing translation for 'onlineSavingsMsg'
Learn More

3-(Difluoromethoxy)aniline, 97%, Thermo Scientific™

Catalog Number p-4258955
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
22236-08-4
C7H7F2NO
159.14
MFCD00236221
RSOFZRXRIPVBBM-UHFFFAOYSA-N
3-difluoromethoxy aniline, m-difluoromethoxyaniline, benzenamine, 3-difluoromethoxy, 3-amino-alpha,alpha-difluoroanisole, 3-difluoromethoxy phenylamine, pubchem8466, acmc-1ckmq, a,a-difluoro-m-anisidine, 3-difluoromethoxyphenylamine
2774110
3-(difluoromethoxy)aniline
NC1=CC(OC(F)F)=CC=C1
This item is not returnable. View return policy
22236-08-4
C7H7F2NO
159.14
MFCD00236221
RSOFZRXRIPVBBM-UHFFFAOYSA-N
3-difluoromethoxy aniline, m-difluoromethoxyaniline, benzenamine, 3-difluoromethoxy, 3-amino-alpha,alpha-difluoroanisole, 3-difluoromethoxy phenylamine, pubchem8466, acmc-1ckmq, a,a-difluoro-m-anisidine, 3-difluoromethoxyphenylamine
2774110
3-(difluoromethoxy)aniline
NC1=CC(OC(F)F)=CC=C1
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 72.0°C to 76.0°C (4.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 159.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-(Difluoromethoxy)aniline
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Difluoromethoxy)aniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'