missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2,5-pyrazinediethanol, TRC

Product Code. 17990785
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990785 2.5 mg 2.5mg
17962145 10 mg 10mg
17901185 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17990785 Supplier Toronto Research Chemicals Supplier No. TRCE9256052.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86917-74-0
Molecular Formula C10 H16 N2 O2
Molecular Weight (g/mol) 196.25
Synonym 2,5-Pyrazinediethanol, 3-ethyl-, 3-Ethyl-2,5-bis(2-hydroxyethyl)pyrazine, Potassium Clavulanate Imp. C (EP)
IUPAC Name 2-[6-ethyl-5-(2-hydroxyethyl)pyrazin-2-yl]ethanol
SMILES CCc1nc(CCO)cnc1CCO

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C10H16N2O2/c1-2-9-10(4-6-14)11-7-8(12-9)3-5-13/h7,13-14H,2-6H2,1H3
Formula Weight 196.1212
Percent Purity >95%
Chemical Name or Material 3-Ethyl-2,5-pyrazinediethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.