Learn More
3'-Fluoro-4'-hydroxyacetophenone, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 403-14-5 |
| Molecular Formula | C8H7FO2 |
| MDL Number | MFCD00215840 |
| Quantity | 5 g |
| SMILES | C1=C(C=CC(=C1F)O)C(C)=O |
| Molecular Weight (g/mol) | 154.14 |
| Percent Purity | 98% |
| Chemical Name or Material | 3'-Fluoro-4'-hydroxyacetophenone |