Learn More
3-Fluoro-4-hydroxybenzoic acid, 98%, Thermo Scientific™
Product Code. 11489687
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489687 5 g 5g
11479687 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11489687 Supplier Thermo Scientific Alfa Aesar Supplier No. B22160.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-fluoro-4- hydroxybenzoic acid is the most potent protocatechuate 3,4-dioxygenase inhibitor.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 350-29-8
Molecular Formula C7H4FO3
Molecular Weight (g/mol) 155.11
MDL Number MFCD00143161
InChI Key IUSDEKNMCOUBEE-UHFFFAOYSA-M
Synonym 4-carboxy-2-fluorophenol, 3-fluoro-4-hydroxy benzoic acid, benzoic acid, 3-fluoro-4-hydroxy, 3-fluoro-4-hydroxybenzoate, 3-fluoro-4-hydroxy-benzoic acid, 3pcf, fhb, pubchem2271, acmc-1co6y, ksc222e7d
PubChem CID 160456
IUPAC Name 3-fluoro-4-hydroxybenzoate
SMILES OC1=CC=C(C=C1F)C([O-])=O
Specifications
Melting Point 157°C to 159°C (decomposition)
Quantity 5 g
Beilstein 2829823
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-hydroxybenzoic acid