Learn More
3-Fluoro-4-iodoaniline, 98%, Thermo Scientific Chemicals
Catalog Number p-7053766
Click to view available options
Quantity:
250 mg
1 g
5 g
656-66-6
C6H5FIN
237.016
MFCD07774187
KUVVJHBHRIXJKI-UHFFFAOYSA-N
3-fluoro-4-iodophenylamine, benzenamine, 3-fluoro-4-iodo, 3-fluoro-4-iodo-aniline, zlchem 541, pubchem1534, 4-iodo 3-fluoro aniline, acmc-1b9hq, 3-fluoro-4-iodo-phenylamine, buttpark 120\07-56
15475546
3-fluoro-4-iodoaniline
C1=CC(=C(C=C1N)F)I
656-66-6
C6H5FIN
237.016
MFCD07774187
KUVVJHBHRIXJKI-UHFFFAOYSA-N
3-fluoro-4-iodophenylamine, benzenamine, 3-fluoro-4-iodo, 3-fluoro-4-iodo-aniline, zlchem 541, pubchem1534, 4-iodo 3-fluoro aniline, acmc-1b9hq, 3-fluoro-4-iodo-phenylamine, buttpark 120\07-56
15475546
3-fluoro-4-iodoaniline
C1=CC(=C(C=C1N)F)I
3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water (228.3 mg/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2811
Sensitivity Light sensitive
Solubility Information Slightly soluble in water (228.3mg/L at 25°C).
Formula Weight 237.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Fluoro-4-iodoaniline