Learn More
3-Hydroxy-2,2,4-trimethyl-pentanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy-2,2,4-trimethyl-pentanoic Acid |
| CAS | 35763-45-2 |
| Molecular Formula | C8H16O3 |
| Quantity | 500 mg |
| Synonym | 3-Hydroxy-2,2,4-trimethyl-valeric Acid, 2,2,4-Trimethyl-3-hydroxyvaleric Acid |
| InChI Formula | InChI=1S/C8H16O3/c1-5(2)6(9)8(3,4)7(10)11/h5-6,9H,1-4H3,(H,10,11) |
| SMILES | CC(C)C(O)C(C)(C)C(=O)O |
| IUPAC Name | 3-hydroxy-2,2,4-trimethylpentanoic acid |
| Molecular Weight (g/mol) | 160.21 |
| Formula Weight | 160.1099 |