Learn More
3-Hydroxy-2,4,6-triiodobenzoic acid, 97%, Thermo Scientific™
Catalog Number p-7077409
Click to view available options
Quantity:
5 g
25 g
100 g
53279-72-4
C7H3I3O3
515.81
MFCD00020305
GIAVHGFPMPSIFI-UHFFFAOYSA-N
htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
96627
3-hydroxy-2,4,6-triiodobenzoic acid
OC(=O)C1=C(I)C=C(I)C(O)=C1I
53279-72-4
C7H3I3O3
515.81
MFCD00020305
GIAVHGFPMPSIFI-UHFFFAOYSA-N
htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
96627
3-hydroxy-2,4,6-triiodobenzoic acid
OC(=O)C1=C(I)C=C(I)C(O)=C1I
Melting Point 211°C to 215°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 515.81
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-2,4,6-triiodobenzoic acid
compliance-icons compliance-icons
Product Identifier
  • 3-Hydroxy-2,4,6-triiodobenzoic acid, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.