Learn More
3-Hydroxy-4-methoxycinnamic Acid, TRC
Product Code. 30134662
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134662 1 g 1g
30133984 10 g 10g
30006157 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134662 Supplier Toronto Research Chemicals Supplier No. TRCH9726901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25522-33-2
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.18
Synonym rans-3-(3-Hydroxy-4-methoxyphenyl)acrylic acid, trans-3-Hydroxy-4-methoxycinnamic acid, Hesperetic acid, trans-Isoferulic acid
IUPAC Name (E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
SMILES COc1ccc(\C=C\C(=O)O)cc1O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C10H10O4/c1-14-9-4-2-7(6-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)/b5-3+
Formula Weight 194.0579
Chemical Name or Material 3-Hydroxy-4-methoxycinnamic Acid