missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxyestra-1,3,5(10)-trien-17beta-yl Butanoate (Estradiol Butyrate), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
3-Hydroxyestra-1,3,5(10)-trien-17beta-yl Butanoate (Estradiol Butyrate)
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxyestra-1,3,5(10)-trien-17beta-yl Butanoate (Estradiol Butyrate) |
| CAS | 18069-79-9 |
| Analyte or Component Names | 3-Hydroxyestra-1,3,5(10)-trien-17beta-yl Butanoate (Estradiol Butyrate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22H30O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3-Hydroxyestra-1,3,5(10)-trien-17beta-yl Butanoate, Estradiol Valerate Imp. F (EP), Estradiol Butyrate, Estradiol Valerate Impurity F |
| SMILES | CCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C |
| Molecular Weight (g/mol) | 342.47 |
| Quantity | 100 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction