Learn More
3-Iodo-1,2-benzenedicarboxylic Acid 1,2-Dimethyl Ester, TRC
Product Code. 30336141
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336141 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336141 Supplier Toronto Research Chemicals Supplier No. TRCB18945550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Iodo-1,2-benzenedicarboxylic Acid 1,2-Dimethyl Ester
CAS 102928-38-1
Molecular Formula C10H9O4I
Quantity 50 mg
InChI Formula InChI=1S/C10H9IO4/c1-14-9(12)6-4-3-5-7(11)8(6)10(13)15-2/h3-5H,1-2H3
SMILES COC(=O)c1cccc(I)c1C(=O)OC
IUPAC Name dimethyl 3-iodobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 320.08
Formula Weight 319.9546