Learn More
3-Iodoanisole, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Iodoanisole |
| CAS | 766-85-8 |
| Molecular Formula | C7H7IO |
| Quantity | 25 g |
| Synonym | 1-Iodo-3-methoxy-benzene, 1-Iodo-3-methoxybenzene, 3-Methoxyphenyl Iodide, m-Methoxyiodobenzene, NSC 60726 |
| InChI Formula | InChI=1S/C7H7IO/c1-9-7-4-2-3-6(8)5-7/h2-5H,1H3 |
| SMILES | COc1cccc(I)c1 |
| IUPAC Name | 1-iodo-3-methoxybenzene |
| Molecular Weight (g/mol) | 234.03 |
| Formula Weight | 233.9542 |
Product Title
Individuate un'opportunità di miglioramento?