missing translation for 'onlineSavingsMsg'
Learn More

3-Iodothiophene, 97%, Thermo Scientific Chemicals

Catalog Number 15224754
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10486-61-0
C4H3IS
210.03
MFCD00037901
WGKRMQIQXMJVFZ-UHFFFAOYSA-N
thiophene, 3-iodo, 3-iodothiophene, stab. with copper, 3-lodothiophene, 3-iodo-thiophene, 3-iodanylthiophene, 3-thienyl iodide, thien-3-yl iodide, pubchem16213, acmc-2098dn
66332
3-iodothiophene
IC1=CSC=C1
This item is not returnable. View return policy
10486-61-0
C4H3IS
210.03
MFCD00037901
WGKRMQIQXMJVFZ-UHFFFAOYSA-N
thiophene, 3-iodo, 3-iodothiophene, stab. with copper, 3-lodothiophene, 3-iodo-thiophene, 3-iodanylthiophene, 3-thienyl iodide, thien-3-yl iodide, pubchem16213, acmc-2098dn
66332
3-iodothiophene
IC1=CSC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -13.0°C
Density 2.0660g/mL
Boiling Point 195.0°C
Flash Point 78°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 2.066
Formula Weight 210.03
Percent Purity 97%
Chemical Name or Material 3-Iodothiophene
Show More Show Less
compliance-icons
Product Identifier
  • 3-Iodothiophene, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'