missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptolactic Acid, TRC

Product Code. 30482446
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482446 1 g 1g
30481263 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30482446 Supplier Toronto Research Chemicals Supplier No. TRCM2534401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2614-83-7
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.14
Synonym 2-Hydroxy-3-mercaptopropanoic Acid, 3-MercaptoLactic Acid, 2-Hydroxy-3-mercaptopropanoic Acid, Hydroxycysteine, β-Mercaptolactic Acid
IUPAC Name 2-hydroxy-3-sulfanylpropanoic acid
SMILES OC(CS)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H6O3S/c4-2(1-7)3(5)6/h2,4,7H,1H2,(H,5,6)
Formula Weight 122.0038
Chemical Name or Material 3-Mercaptolactic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.