missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-1-propanol, 98%, Thermo Scientific Chemicals

Catalog Number p-4259005
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1589-49-7
C4H10O2
90.12
JDFDHBSESGTDAL-UHFFFAOYSA-N
3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol
74116
3-methoxypropan-1-ol
COCCCO
This item is not returnable. View return policy
1589-49-7
C4H10O2
90.12
JDFDHBSESGTDAL-UHFFFAOYSA-N
3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol
74116
3-methoxypropan-1-ol
COCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9420g/mL
Boiling Point 149.0°C to 150.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3O(CH2)3OH
Refractive Index 1.4120 to 1.4140 (20°C, 589nm)
Quantity 10 g
Specific Gravity 0.942
Formula Weight 90.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1-propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methoxy-1-propanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P233-Keep container tightly closed.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P242-Use only non-sparking tools.
  • P240-Ground/bond container and receiving equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'