missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-phenyl-4H-furo[2,3-h]chromen-4-one, TRC

Product Code. 30482222
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482222 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482222 Supplier Toronto Research Chemicals Supplier No. TRCM078060250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Karanjin
CAS 521-88-0
Molecular Formula C18 H12 O4
Quantity 250 mg
Synonym Karanjin (6CI), 3-Methoxy-2-phenyl-4H-furo[2,3-h]-1-benzopyran-4-one, Derisom, NSC 335755, Karanjin
InChI Formula InChI=1S/C18H12O4/c1-20-18-15(19)13-7-8-14-12(9-10-21-14)17(13)22-16(18)11-5-3-2-4-6-11/h2-10H,1H3
SMILES COC1=C(OC2=C(C1=O)C=CC3=C2C=CO3)C4=CC=CC=C4
IUPAC Name 3-methoxy-2-phenylfuro[2,3-h]chromen-4-one
Molecular Weight (g/mol) 292.29
Formula Weight 292.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.