Learn More
3-Methoxy-3-methylbutan-1-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methoxy-3-methylbutan-1-ol |
| CAS | 56539-66-3 |
| Molecular Formula | C6H14O2 |
| Quantity | 1 g |
| Synonym | Finetop J 210, 3-Methyl-3-(methyloxy)butan-1-ol, Solfit MMB |
| InChI Formula | InChI=1S/C6H14O2/c1-6(2,8-3)4-5-7/h7H,4-5H2,1-3H3 |
| SMILES | COC(C)(C)CCO |
| IUPAC Name | 3-methoxy-3-methylbutan-1-ol |
| Molecular Weight (g/mol) | 118.17 |
| Formula Weight | 118.0994 |