missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxytyramine, MedChemExpress

Product Code. 16424926
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
Product Code. Quantity unitSize
16424926 100 mg 100mg
16461576 500 mg 500mg
16429855 250 mg 250mg
3 options
This item is not returnable. View return policy

Product Code. 16424926

Brand: Medchem Express HY103638A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Methoxytyramine, a well known extracellular metabolite of 3-hydroxytyramine/dopamine, is a neuromodulator.

3-Methoxytyramine, a well known extracellular metabolite of 3-hydroxytyramine/dopamine, is a neuromodulator.

Specifications

Chemical Name or Material 3-Methoxytyramine
CAS 554-52-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C9H13NO2
Quantity 100 mg
Synonym 3-O-methyl Dopamine
Solubility Information DMSO : 8.33 mg/mL (49.82 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2 mg/mL (11.96 mM; Need ultrasonic)
SMILES OC1=CC=C(CCN)C=C1OC
Molecular Weight (g/mol) 167.21
Formula Weight 167.21
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.