missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxytyramine, MedChemExpress
Description
3-Methoxytyramine, a well known extracellular metabolite of 3-hydroxytyramine/dopamine, is a neuromodulator.
Specifications
Specifications
| Chemical Name or Material | 3-Methoxytyramine |
| CAS | 554-52-9 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C9H13NO2 |
| Quantity | 500 mg |
| Synonym | 3-O-methyl Dopamine |
| Solubility Information | DMSO : 8.33 mg/mL (49.82 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2 mg/mL (11.96 mM; Need ultrasonic) |
| SMILES | OC1=CC=C(CCN)C=C1OC |
| Molecular Weight (g/mol) | 167.21 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?