missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-[(5-phenylpentanoyl)[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
3-Methyl-2-[(5-phenylpentanoyl)[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic Acid
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-2-[(5-phenylpentanoyl)[(2'-(1H-tetrazol-5-yl)biphenyl-4-yl)methyl]amino]butanoic Acid |
| Analyte or Component Names | 3-Methyl-2-[(5-phenylpentanoyl)[(2'-(1H-tetrazol-5-yl)biphenyl-4-yl)methyl]amino]butanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H33 N5 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Methyl-2-[(5-phenylpentanoyl)[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic Acid |
| InChI Formula | InChI=1S/C30H33N5O3/c1-21(2)28(30(37)38)35(27(36)15-9-6-12-22-10-4-3-5-11-22)20-23-16-18-24(19-17-23)25-13-7-8-14-26(25)29-31-33-34-32-29/h3-5,7-8,10-11,13-14,16-19,21,28H,6,9,12,15,20H2,1-2H3,(H,37,38)(H,31,32,33,34) |
| SMILES | CC(C)C(N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)C(=O)CCCCc4ccccc4)C(=O)O |
| IUPAC Name | 3-methyl-2-[5-phenylpentanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]butanoic acid |
| Molecular Weight (g/mol) | 511.6147 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction