missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-butanone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1000 mL
Description
3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.
Solubility
Soluble in water (8.21 mg/ml at 20°C), ether, alcohol, acetone, and ethanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.
Solubility
Soluble in water (8.21 mg/ml at 20°C), ether, alcohol, acetone, and ethanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Melting Point | -92°C |
Density | 0.805 |
Boiling Point | 94°C to 95°C |
Flash Point | −1°C (30°F) |
Odor | Fruit-like |
Refractive Index | 1.388 |
Quantity | 250 mL |
UN Number | UN2397 |
Beilstein | 1071238 |
Merck Index | 14,6090 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'