missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-[propanoyl[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
3-Methyl-2-[propanoyl[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic Acid
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-2-[propanoyl[(2'-(1H-tetrazol-5-yl)biphenyl-4-yl)methyl]amino]butanoic Acid |
| Analyte or Component Names | 3-Methyl-2-[propanoyl[(2'-(1H-tetrazol-5-yl)biphenyl-4-yl)methyl]amino]butanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H25 N5 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Methyl-2-[propanoyl[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic Acid |
| InChI Formula | InChI=1S/C22H25N5O3/c1-4-19(28)27(20(14(2)3)22(29)30)13-15-9-11-16(12-10-15)17-7-5-6-8-18(17)21-23-25-26-24-21/h5-12,14,20H,4,13H2,1-3H3,(H,29,30)(H,23,24,25,26) |
| SMILES | CCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)C(C(C)C)C(=O)O |
| IUPAC Name | 3-methyl-2-[propanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]butanoic acid |
| Molecular Weight (g/mol) | 407.4656 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction