missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-(tri-n-butylstannyl)pyridine, 96%

Product Code. 15408127
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408127 250 mg 250mg
15498117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408127 Supplier Thermo Scientific Alfa Aesar Supplier No. H51540.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 259807-97-1 | C18H33NSn | 382.179 g/mol

3-Methyl-2-(tri-n-butylstannyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-(tri-n-butylstannyl)pyridine is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259807-97-1
Molecular Formula C18H33NSn
Molecular Weight (g/mol) 382.179
MDL Number MFCD07787404
InChI Key QWDWZQDTTVYOBK-UHFFFAOYSA-N
Synonym 3-methyl-2-tributylstannyl pyridine, 2-tributylstannyl-3-methylpyridine, pyridine, 3-methyl-2-tributylstannyl, 3-methyl-2-tri-n-butylstannyl pyridine, acmc-209gnt, 3-methyl-2-tributylstannanylpyridine, 2-tri-n-butylstannyl-3-methylpyridine
PubChem CID 16427098
IUPAC Name tributyl-(3-methylpyridin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=C(C=CC=N1)C

Specifications

Quantity 250 mg
UN Number UN2788
Solubility Information Sparingly soluble in water.
Formula Weight 382.16
Percent Purity 96%
Chemical Name or Material 3-Methyl-2-(tri-n-butylstannyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.