missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-3-octanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Density | 0.8220g/mL |
Boiling Point | 127.0°C |
Flash Point | 72°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Specific Gravity | 0.822 |
Formula Weight | 144.26 |
Percent Purity | 97% |
Show More |
Safety and Handling
Product Identifier
- 3-Methyl-3-octanol
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'