missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzotrifluoride, 97%, Thermo Scientific™

Catalog Number p-4380097
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
67192-42-1
PCCXJSOGGOWRJC-UHFFFAOYSA-N
3-methyl-4-nitrobenzotrifluoride, 2-methyl-1-nitro-4-trifluoromethyl benzene, 4-trifluoromethyl-2-methyl-1-nitrobenzene, 2-methyl-1-nitro-4-trifluoromethyl-benzene, 3-methyl-4-nitrobenzotrifluoroide, 1-nitro-2-methyl-4-trifluoromethyl benzene, benzene, 2-methyl-1-nitro-4-trifluoromethyl
144223
2-methyl-1-nitro-4-(trifluoromethyl)benzene
CC1=C(C=CC(=C1)C(F)(F)F)[N+](=O)[O-]
This item is not returnable. View return policy
67192-42-1
PCCXJSOGGOWRJC-UHFFFAOYSA-N
3-methyl-4-nitrobenzotrifluoride, 2-methyl-1-nitro-4-trifluoromethyl benzene, 4-trifluoromethyl-2-methyl-1-nitrobenzene, 2-methyl-1-nitro-4-trifluoromethyl-benzene, 3-methyl-4-nitrobenzotrifluoroide, 1-nitro-2-methyl-4-trifluoromethyl benzene, benzene, 2-methyl-1-nitro-4-trifluoromethyl
144223
2-methyl-1-nitro-4-(trifluoromethyl)benzene
CC1=C(C=CC(=C1)C(F)(F)F)[N+](=O)[O-]
Boiling Point 95.0°C (13.0 Torr)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H6F3NO2
Quantity 1 g
Molecular Weight (g/mol) 205.13
Formula Weight 205.13
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzotrifluoride
compliance-icons
Product Identifier
  • 3-Methyl-4-nitrobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'