Learn More
3-(Methylamino)-1-phenylpropan-1-one Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Methylamino)-1-phenylpropan-1-one Hydrochloride |
| CAS | 2538-50-3 |
| Molecular Formula | C10H13NO . HCl |
| Quantity | 100 mg |
| Synonym | 3-(N-Methylamino)propiophenone Hydrochloride |
| InChI Formula | InChI=1S/C10H13NO.ClH/c1-11-8-7-10(12)9-5-3-2-4-6-9;/h2-6,11H,7-8H2,1H3;1H |
| SMILES | Cl.CNCCC(=O)c1ccccc1 |
| IUPAC Name | 3-(methylamino)-1-phenylpropan-1-one;hydrochloride |
| Molecular Weight (g/mol) | 199.68 |
| Formula Weight | 199.0764 |