Learn More
3-Methylflavone-8-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methylflavone-8-Carboxylic Acid |
| CAS | 3468-01-7 |
| Molecular Formula | C17 H12 O4 |
| Quantity | 10 g |
| Synonym | 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic Acid, Flavoxate Hydrochloride Imp. A (EP), 3-Methyl-flavone-8-carboxylic Acid |
| InChI Formula | InChI=1S/C17H12O4/c1-10-14(18)12-8-5-9-13(17(19)20)16(12)21-15(10)11-6-3-2-4-7-11/h2-9H,1H3,(H,19,20) |
| SMILES | CC1=C(Oc2c(cccc2C1=O)C(=O)O)c3ccccc3 |
| IUPAC Name | 3-methyl-4-oxo-2-phenylchromene-8-carboxylic acid |
| Molecular Weight (g/mol) | 280.27 |
| Formula Weight | 280.0736 |