missing translation for 'onlineSavingsMsg'
Learn More

3-(N-BOC)-(2-aminoethyl)azetidine, 96%, Thermo Scientific™

Catalog Number 15249696
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
162696-31-3
C10H20N2O2
200.28
CSPJUZSNJFANNU-UHFFFAOYSA-N
3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl n-2-azetidin-3-yl ethyl carbamate, 3-2-n-boc-aminoethyl azetidine, tert-butyl 2-3-azetidinyl ethylcarbamate, 2-azetin-3-yl-1-n-boc ethylamine, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester
37818431
tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
CC(C)(C)OC(=O)NCCC1CNC1
This item is not returnable. View return policy
162696-31-3
C10H20N2O2
200.28
CSPJUZSNJFANNU-UHFFFAOYSA-N
3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl n-2-azetidin-3-yl ethyl carbamate, 3-2-n-boc-aminoethyl azetidine, tert-butyl 2-3-azetidinyl ethylcarbamate, 2-azetin-3-yl-1-n-boc ethylamine, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester
37818431
tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
CC(C)(C)OC(=O)NCCC1CNC1
Melting Point 84°C to 86°C
Assay Percent Range 95% min. (HPLC)
Formula Weight 200.28
Percent Purity 96%
Quantity 250 mg
Chemical Name or Material 3-(N-BOC)-(2-aminoethyl)azetidine
compliance-icons
Product Identifier
  • 3-(N-Boc)-(2-aminoethyl)azetidine, 96%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'