missing translation for 'onlineSavingsMsg'
Learn More

3-n-Octylthiophene, 97%, Thermo Scientific Chemicals

Catalog Number p-7052715
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
65016-62-8
C12H20S
196.352
MFCD00085281
WQYWXQCOYRZFAV-UHFFFAOYSA-N
3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
566852
3-octylthiophene
CCCCCCCCC1=CSC=C1
This item is not returnable. View return policy
65016-62-8
C12H20S
196.352
MFCD00085281
WQYWXQCOYRZFAV-UHFFFAOYSA-N
3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
566852
3-octylthiophene
CCCCCCCCC1=CSC=C1

3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).

Solubility
Soluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.92 g/mL
Boiling Point 106°C to 107°C (3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.492
Quantity 1 g
Solubility Information Soluble in organic solvents.
Formula Weight 196.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-n-Octylthiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'