Learn More
Eppendorf™ Research 3 Neo Factory Adjustment Seal
Shop All Eppendorf Products
Click to view available options
Type:
Protective Seal
Unit Size:
Pack of 10
Description
- Factory Adjustment Seal specifically designed for Eppendorf Research™ 3 Neo pipettes
- Constructed from high-quality Polypropylene for durability and chemical resistance
- Acts as a protective seal during factory calibration or pipette adjustments
- Ensures tamper-evidence and maintains instrument integrity
- Ideal for maintaining accuracy and compliance in laboratory environments
Specifications
Specifications
| For Use With | Pipettes |
| Material | Polypropylene |
| Type | Protective Seal |
| For Use With (Equipment) | 3 Neo, Adjustment Seal |
| Quantity | 1 Pk. |