Learn More
Eppendorf™ ColorTag Marking Rings - Sample Bag
Product Code. 30689742 Shop All Eppendorf Products
Click to view available options
Unit Size:
Pack of 5
Product Code. 30689742
Brand: Eppendorf™ 3102666000
Autoclavable and suitable for UV or H2O2 sterilization as well as extensively tested for lab use
  • Flexible silicone rings for easy pipette and dispenser identification and labelling
  • Label any pipette for personal use or specific applications (6 sizes, writable with markers)
  • Prevent errors from pipette misidentification and add color and joy to your lab (12 vibrant colors)
  • Autoclavable and suitable for UV or H2O2 sterilization as well as extensively tested for lab use
  • ColorTag rings have the Eppendorf logo embossed inside for easy recognition of the original
  • Eco-friendly, re-closable packaging made from 100 % paper
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Silicone
For Use With (Equipment) 6 sizes, Writable With Markers
Includes ColorTag, Sample pack
Quantity 12 Pk.