Learn More
3-Nitro-4-hydroxyquinoline, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Nitro-4-hydroxyquinoline |
| CAS | 50332-66-6 |
| Molecular Formula | C9 H6 N2 O3 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C9H6N2O3/c12-9-6-3-1-2-4-7(6)10-5-8(9)11(13)14/h1-5H,(H,10,12) |
| SMILES | Oc1c(cnc2ccccc12)[N+](=O)[O-] |
| IUPAC Name | 3-nitroquinolin-4-ol |
| Molecular Weight (g/mol) | 190.16 |
| Formula Weight | 190.0378 |