Learn More
3-Nitrophenyl isothiocyanate, 97%
Product Code. 11357716
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357716 25 g 25g
11337716 1 g 1g
11347716 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11357716 Supplier Thermo Scientific Alfa Aesar Supplier No. L11335.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 3529-82-6 | C7H4N2O2S | 180.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3529-82-6
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.18
MDL Number MFCD00041245
InChI Key OEZXLKSZOAWNJU-UHFFFAOYSA-N
Synonym 3-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-3-nitro, m-nitrophenyl isothiocyanate, 3-nitrophenylisothiocyanate, isothiocyanic acid, m-nitrophenyl ester, 1-isothiocyanato-3-nitro-benzene, m-nitrophenylisothiocyanat, acmc-1co3y, phenyl, 3-nitro, isothiocyanate, #
PubChem CID 19050
IUPAC Name 1-isothiocyanato-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(=C1)N=C=S
Specifications
Melting Point 56°C to 58°C
Odor Irritating
Quantity 25 g
UN Number UN2923
Beilstein 2094061
Sensitivity Moisture sensitive
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 3-Nitrophenyl isothiocyanate