Learn More
3-Oxo-4-aza-5Alpha-Alphandrost-1-ene-17Beta-carboxylic Acid Methyl Ester, TRC
Product Code. 30136495
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136495 100 mg 100mg
30003925 25 mg 25mg
17920355 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136495 Supplier Toronto Research Chemicals Supplier No. TRCO847260100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103335-41-7
Molecular Formula C20 H29 N O3
Molecular Weight (g/mol) 331.45
Synonym Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate, Finasteride Imp. B (EP), Delta-1-Aza Ester of Finasteride, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, methyl ester, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxylic acid, 3-oxo-, methyl ester, (5α,17β)-, 17β-(Methoxycarbonyl)-4-aza-5α-androst-1-en-3-one, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 1,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, methyl ester, [4aR-(4aα,4bβ,6aα,7α,9aβ,9bα,11aβ)]-, 3-Oxo-4-aza-5α-androst-1-en-17β-carboxylic acid methyl ester, Methyl 3-oxo-4-aza-5α-androst-1-ene-17β-carboxylate
IUPAC Name methyl (1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxylate
SMILES COC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C20H29NO3/c1-19-10-8-14-12(13(19)5-6-15(19)18(23)24-3)4-7-16-20(14,2)11-9-17(22)21-16/h9,11-16H,4-8,10H2,1-3H3,(H,21,22)/t12-,13-,14-,15+,16+,19-,20+/m0/s1
Formula Weight 331.2147
Percent Purity >95%
Chemical Name or Material Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate (delta-1-Aza Ester)
Grade HPLC