Learn More
3-Oxotetrahydrofuran, TRC
Product Code. 30417470
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417470 500 mg -
30423489 1 g -
30425853 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30417470 Supplier Toronto Research Chemicals Supplier No. TRCO870480500MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Denne vare kan ikke returneres. Se returpolicy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22929-52-8
Molecular Formula C4 H6 O2
Molecular Weight (g/mol) 86.09
Synonym 4,5-Dihydro-3(2H)-furanone, Dihydro-3(2H)-furanone, Dihydrofuran-3-one, Oxolan-3-one
IUPAC Name oxolan-3-one
SMILES O=C1CCOC1
Tekniske data
Quantity 500 mg
InChI Formula InChI=1S/C4H6O2/c5-4-1-2-6-3-4/h1-3H2
Formula Weight 86.0368
Chemical Name or Material 3-Oxotetrahydrofuran
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.