missing translation for 'onlineSavingsMsg'
Learn More

3-Oxotetrahydrofuran, TRC

Product Code. 30423489
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30423489 1 g -
30425853 5 g 5g
30417470 500 mg -
3 options
This item is not returnable. View return policy

Product Code. 30423489

Brand: Toronto Research Chemicals TRCO8704801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22929-52-8
Formula Weight 86.0368
InChI Formula InChI=1S/C4H6O2/c5-4-1-2-6-3-4/h1-3H2
IUPAC Name oxolan-3-one
Molecular Formula C4 H6 O2
Molecular Weight (g/mol) 86.09
SMILES O=C1CCOC1
Synonym 4,5-Dihydro-3(2H)-furanone, Dihydro-3(2H)-furanone, Dihydrofuran-3-one, Oxolan-3-one
Chemical Name or Material 3-Oxotetrahydrofuran

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.