Learn More
3-Phenanthrenecarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Phenanthrenecarboxylic Acid |
| CAS | 7470-14-6 |
| Molecular Formula | C15H10O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C15H10O2/c16-15(17)12-8-7-11-6-5-10-3-1-2-4-13(10)14(11)9-12/h1-9H,(H,16,17) |
| SMILES | O=C(O)C1=CC2=C(C=C1)C=CC3=C2C=CC=C3 |
| IUPAC Name | phenanthrene-3-carboxylic acid |
| Molecular Weight (g/mol) | 222.24 |
| Formula Weight | 222.0681 |