Learn More
3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)prop-2-enenitrile (E/Z-mixture), Mikromol™
Description
3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)prop-2-enenitrile (E/Z-mixture)
Specifications
Specifications
| Chemical Name or Material | 3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)prop-2-enenitrile (E,Z mixture) |
| CAS | 30078-48-9 |
| Analyte or Component Names | 3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)prop-2-enenitrile (E,Z mixture) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H20 N2 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Trimethoprim Imp. I (EP), 3-Anilino-2-(3,4,5-trimethoxybenzyl) acrylonitrile, 3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)prop-2-enenitrile (E/Z-mixture), Trimethoprim Impurity I |
| InChI Formula | InChI=1S/C19H20N2O3/c1-22-17-10-14(11-18(23-2)19(17)24-3)9-15(12-20)13-21-16-7-5-4-6-8-16/h4-8,10-11,13,21H,9H2,1-3H3 |
| SMILES | COc1cc(CC(=CNc2ccccc2)C#N)cc(OC)c1OC |
| IUPAC Name | 3-anilino-2-[(3,4,5-trimethoxyphenyl)methyl]prop-2-enenitrile |
| Show More |