missing translation for 'onlineSavingsMsg'
Learn More

3-Picolyl chloride hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-3779471
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6959-48-4
C6H6ClN·HCl
164.04
MFCD00012818
UZGLOGCJCWBBIV-UHFFFAOYSA-N
3-chloromethyl pyridine hydrochloride, 3-chloromethylpyridine hydrochloride, 3-picolyl chloride hydrochloride, 3-chloromethyl pyridine hcl, ccris 140, pyridine, 3-chloromethyl-, hydrochloride, 3-pyridylmethyl chloride hydrochloride, unii-bqa8fyp4yn, 3-chloromethyl pyridinium chloride, 3-chloromethyl pyridine.hcl
23394
3-(chloromethyl)pyridine;hydrochloride
C1=CC(=CN=C1)CCl.Cl
This item is not returnable. View return policy
6959-48-4
C6H6ClN·HCl
164.04
MFCD00012818
UZGLOGCJCWBBIV-UHFFFAOYSA-N
3-chloromethyl pyridine hydrochloride, 3-chloromethylpyridine hydrochloride, 3-picolyl chloride hydrochloride, 3-chloromethyl pyridine hcl, ccris 140, pyridine, 3-chloromethyl-, hydrochloride, 3-pyridylmethyl chloride hydrochloride, unii-bqa8fyp4yn, 3-chloromethyl pyridinium chloride, 3-chloromethyl pyridine.hcl
23394
3-(chloromethyl)pyridine;hydrochloride
C1=CC(=CN=C1)CCl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 137°C to 143°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 820g/L
Formula Weight 164.04
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 3-Picolyl chloride hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'