missing translation for 'onlineSavingsMsg'
Learn More

3-Propionylpyridine, 98%, Thermo Scientific™

Catalog Number p-7077502
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
This item is not returnable. View return policy
1570-48-5
C8H9NO
135.17
VDNKJMUNLKAGAM-UHFFFAOYSA-N
3-propionylpyridine, 1-3-pyridyl propan-1-one, ethyl 3-pyridyl ketone, 1-pyridin-3-yl propan-1-one, 3-pyridyl ethyl ketone, 1-3-pyridinyl-1-propanone, 3-propionyl pyridine, 1-pyridin-3-yl-propan-1-one, ethyl pyridin-3-yl ketone, 1-propanone, 1-3-pyridinyl
74074
1-pyridin-3-ylpropan-1-one
CCC(=O)C1=CN=CC=C1
This item is not returnable. View return policy
1570-48-5
C8H9NO
135.17
VDNKJMUNLKAGAM-UHFFFAOYSA-N
3-propionylpyridine, 1-3-pyridyl propan-1-one, ethyl 3-pyridyl ketone, 1-pyridin-3-yl propan-1-one, 3-pyridyl ethyl ketone, 1-3-pyridinyl-1-propanone, 3-propionyl pyridine, 1-pyridin-3-yl-propan-1-one, ethyl pyridin-3-yl ketone, 1-propanone, 1-3-pyridinyl
74074
1-pyridin-3-ylpropan-1-one
CCC(=O)C1=CN=CC=C1
Boiling Point 106.0°C (3.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material 3-Propionylpyridine
compliance-icons
Product Identifier
  • 3-Propionylpyridine, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'