missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7077387
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
175277-11-9
C9H14N2O2
182.22
SFSXXMXHJOSBAZ-UHFFFAOYSA-N
3-tert-butyl-1-methyl-1h-pyrazole-5-carboxylic acid, 3-tert-butyl-1-methylpyrazole-5-carboxylic acid, 5-tert-butyl-2-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 3-1,1-dimethylethyl-1-methyl, 3-t-butyl-1-methyl-1h-pyrazole-5-carboxylic acid, maybridge1_008671, acmc-1bzn5, butylmethylpyrazolecarboxylicacid
720529
5-tert-butyl-2-methylpyrazole-3-carboxylic acid
CC(C)(C)C1=NN(C(=C1)C(=O)O)C
This item is not returnable. View return policy
175277-11-9
C9H14N2O2
182.22
SFSXXMXHJOSBAZ-UHFFFAOYSA-N
3-tert-butyl-1-methyl-1h-pyrazole-5-carboxylic acid, 3-tert-butyl-1-methylpyrazole-5-carboxylic acid, 5-tert-butyl-2-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 3-1,1-dimethylethyl-1-methyl, 3-t-butyl-1-methyl-1h-pyrazole-5-carboxylic acid, maybridge1_008671, acmc-1bzn5, butylmethylpyrazolecarboxylicacid
720529
5-tert-butyl-2-methylpyrazole-3-carboxylic acid
CC(C)(C)C1=NN(C(=C1)C(=O)O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 151.0°C to 154.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 182.22
Percent Purity 95%
Chemical Name or Material 3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid
compliance-icons
Product Identifier
  • 3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'