missing translation for 'onlineSavingsMsg'
Learn More

3-[(tert-Butyldimethylsilyl)oxy]-1-propanal, TRC

Product Code. 30346512
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346512 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346512 Supplier Toronto Research Chemicals Supplier No. TRCB6915651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[(tert-Butyldimethylsilyl)oxy]-1-propanal
CAS 89922-82-7
Molecular Formula C9 H20 O2 Si
Quantity 1 g
InChI Formula InChI=1S/C9H20O2Si/c1-9(2,3)12(4,5)11-8-6-7-10/h7H,6,8H2,1-5H3
SMILES CC(C)(C)[Si](C)(C)OCCC=O
IUPAC Name 3-[tert-butyl(dimethyl)silyl]oxypropanal
Molecular Weight (g/mol) 188.34
Formula Weight 188.1233
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.