missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number 15224408
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
220227-84-9
C7H4ClF3O3S
260.62
DODDSXTWDSJCDN-UHFFFAOYSA-N
3-trifluoromethoxy benzenesulfonyl chloride, 3-trifluoromethoxy benzene-1-sulfonyl chloride, 3-trifluoromethoxy benzenesulphonyl chloride, 3-trifluoromethoxy-benzenesulfonyl chloride, 3-trifluoromethoxybenzenesulfonylchloride, 3-trifluoromethyl oxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3-trifluoromethoxy, pubchem5187, acmc-1ce4e
2777218
3-(trifluoromethoxy)benzenesulfonyl chloride
C1=CC(=CC(=C1)S(=O)(=O)Cl)OC(F)(F)F
This item is not returnable. View return policy
220227-84-9
C7H4ClF3O3S
260.62
DODDSXTWDSJCDN-UHFFFAOYSA-N
3-trifluoromethoxy benzenesulfonyl chloride, 3-trifluoromethoxy benzene-1-sulfonyl chloride, 3-trifluoromethoxy benzenesulphonyl chloride, 3-trifluoromethoxy-benzenesulfonyl chloride, 3-trifluoromethoxybenzenesulfonylchloride, 3-trifluoromethyl oxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3-trifluoromethoxy, pubchem5187, acmc-1ce4e
2777218
3-(trifluoromethoxy)benzenesulfonyl chloride
C1=CC(=CC(=C1)S(=O)(=O)Cl)OC(F)(F)F
Boiling Point 73.0°C to 74.0°C (0.7 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 260.62
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethoxy)benzenesulfonyl chloride
compliance-icons
Product Identifier
  • 3-(Trifluoromethoxy)benzenesulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'