Learn More
3-(Trifluoromethyl)anisole, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 454-90-0 |
| Molecular Formula | C8H7F3O |
| MDL Number | MFCD00000385 |
| Quantity | 25 g |
| SMILES | C1=C(C=CC=C1OC)C(F)(F)F |
| Molecular Weight (g/mol) | 176.14 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-(Trifluoromethyl)anisole |