missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)pyridine-4-carboxylic acid, 95%

Product Code. 15409728
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409728 5 g 5g
15448166 250 mg 250mg
15499718 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15409728 Supplier Thermo Scientific Alfa Aesar Supplier No. H63174.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590371-38-3 | C7H4F3NO2 | 191.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590371-38-3
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD08234940
InChI Key FBOSFEOQEZQFOC-UHFFFAOYSA-N
Synonym 3-trifluoromethyl pyridine-4-carboxylic acid, 3-trifluoromethyl isonicotinic acid, 3-trifluoromethyl-isonicotinic acid, 3-trifluoromethyl-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-trifluoromethyl, pubchem22216, ksc268s9d, 3-trifluoromethylisonicotinic acid, 3-trifluoromethyl isonicotinicacid
PubChem CID 25210500
IUPAC Name 3-(trifluoromethyl)pyridine-4-carboxylic acid
SMILES OC(=O)C1=C(C=NC=C1)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 191.11
Percent Purity 95%
Chemical Name or Material 3-(Trifluoromethyl)pyridine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.