Learn More
3-(Trifluoromethyl)pyridine-4-carboxylic acid, 95%
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 191.11 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-(Trifluoromethyl)pyridine-4-carboxylic acid |