Learn More
3-(Trifluoromethylthio)aniline, 96%
Product Code. p-7027179
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11400168 1 g 1g
11410168 5 g 5g
2 options
Product Code. 11400168
Brand: Thermo Scientific Alfa Aesar B23734.03
Request Bulk or Custom Format
CAS: 369-68-6 | C7H6F3NS | 193.19 g/mol
3-(Trifluoromethylthio)aniline is used as an intermediate in organic synthesis. It is also involved in the regioselective Friedel-Crafts alkylation in hexafluoroacetone sesquihydrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(Trifluoromethylthio)aniline is used as an intermediate in organic synthesis. It is also involved in the regioselective Friedel-Crafts alkylation in hexafluoroacetone sesquihydrate.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 369-68-6
Molecular Formula C7H6F3NS
Molecular Weight (g/mol) 193.19
MDL Number MFCD00190130
InChI Key DENPAKQJZNDKEL-UHFFFAOYSA-N
Synonym 3-trifluoromethylthio aniline, 3-trifluoromethyl sulfanyl aniline, 3-aminophenyl trifluoromethyl sulphide, 3-aminophenyl trifluoromethyl sulfide, 3-trifluoromethyl thio aniline, 3-trifluoromethylsulfanyl aniline, benzenamine, 3-trifluoromethyl thio, benzeneamine, 3-trifluoromethyl thio, 3-trifluoromethylthio phenylamine
PubChem CID 2735939
IUPAC Name 3-(trifluoromethylsulfanyl)aniline
SMILES NC1=CC=CC(SC(F)(F)F)=C1
Specifications
Density 1.348
Boiling Point 221°C to 224°C
Flash Point 103°C (218°F)
Odor Unpleasant
Refractive Index 1.522
Quantity 1 g
UN Number UN2810
Solubility Information Immiscible with water.
Formula Weight 193.19
Percent Purity 96%
Chemical Name or Material 3-(Trifluoromethylthio)aniline