Learn More
[3-(Trimethylammonium)propyl] Methanethiosulfonate Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | (3-(Trimethylammonium)propyl) Methanethiosulfonate Bromide |
| CAS | 220560-60-1 |
| Molecular Formula | C7 H18 N O2 S2 . Br |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C7H18NO2S2.BrH/c1-8(2,3)6-5-7-11-12(4,9)10;/h5-7H2,1-4H3;1H/q+1;/p-1 |
| SMILES | [Br-].C[N+](C)(C)CCCSS(=O)(=O)C |
| IUPAC Name | trimethyl(3-methylsulfonylsulfanylpropyl)azanium;bromide |
| Molecular Weight (g/mol) | 292.26 |
| Formula Weight | 290.9962 |