Learn More
Watson-Marlow Pumps 300 Series Drive
Click to view available options
Model:
323Du
323Dz
323E
323S
323U
Unit Size:
Each
Description
- 300 series laboratory pumps are a benchtop pumping solution with manual, remote, analog or RS232 control capabilities.
- Featuring a chemically resistant ABS top and IP31 protection, 323 pumps have a membrane keypad that controls the pump, and a digital LCD to display speed.
- There are four models with increasing degrees of functionality:
- 323Du RS232/auto/manual enhanced-control high-flow pumps
- 323U and 323U analogue-auto and enhanced manual-control pumps
- 323S high-performance manual control pumps
- 323E manual control pumps
Pumphead options available as standard (others available on request):
- 313D and 313D2 three-roller pumpheads for continuous tubing
- 314D and 314D2 four-roller pumpheads for continuous tubing
- 314MC four-roller five-channel microcassette pumpheads
- 318MC eight-roller five-channel microcassette pumpheads
- 501RL and 501RL2 pumpheads for continuous tubing
Specifications
Specifications
| Display | LCD |
| Frequency | 50/60 Hz |
| IP Rating | IP31 |
| Material | ABS Plastic, Aluminum |
| Model | 323Dz |
| Plug Type | UK |
| Type | Cased Pump |
| Voltage | 100/120/220/240 V |
| Speed | 400 rpm |
| Case Material | ABS, Powder Coated Aluminium LM24 |