missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetoacetyl Coenzyme A Sodium Hydrate (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | Acetoacetyl Coenzyme A Sodium Salt Hydrate |
| Analyte or Component Names | Acetoacetyl Coenzyme A Sodium Salt Hydrate |
| Shipping Condition | Blue Ice Dry Pack |
| Molecular Formula | C25 H40 N7 O18 P3 S . xNa . xH2O |
| InChI Formula | InChI=1S/C25H40N7O18P3S.Na.H2O/c1-13(33)8-16(35)54-7-6-27-15(34)4-5-28-23(38)20(37)25(2,3)10-47-53(44,45)50-52(42,43)46-9-14-19(49-51(39,40)41)18(36)24(48-14)32-12-31-17-21(26)29-11-30-22(17)32;;/h11-12,14,18-20,24,36-37H,4-10H2,1-3H3,(H,27,34)(H,28,38)(H,42,43)(H,44,45)(H2,26,29,30)(H2,39,40,41);;1H2/q;+1;/t14-,18-,19-,20+,24-;;/m1../s1 |
| Solution Type | Neat |
| SMILES | O.[Na+].CC(=O)CC(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP(=O)(O)OP(=O)(O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(=O)(O)O)n2cnc3c(N)ncnc23 |
| IUPAC Name | sodium;S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxy-tetrahydrofuran-2-yl]methoxy-hydroxy-phosphoryl]oxy-hydroxy-phosphoryl]oxy-2-hydroxy-3,3-dimethyl-butanoyl]amino]propanoylamino]ethyl] 3-oxobutanethioate;hydrate |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 892.6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?